Learn More
Essendant GLUEEPOXY 5 MIN SET
Supplier:  Essendant LOC1365868
Encompass_Preferred
Easy-to-use self-mixing dispenser and precision applicator High-strength formula bonds wood metal tile ceramic glass plastic and more Final bond is high strength and weather- water- and solvent-resistant Sandable and paintable Adhesive Type- Liquid Product Packaging- Tube Washable/Removable- Yes Color on Application- White
Catalog No. 50-000-31815