Learn More
Essendant GLUELOCTITE ULTRA GEL4G
Supplier:  Essendant LOC1363589
Encompass_Preferred
Features rubber-infused technology which creates a flexible bond that resists impact twisting and pulling Works in all temperatures and conditions Gel formula does not run and the squeeze side grips make dispensing the right amount of glue a snap Adhesive Type- Gel Product Packaging- Bottle Washable/Removable- Yes Color on Application- White
Catalog No. 50-000-31814