Learn More
Essendant GLUESTICKDUAL MELT24PK
Supplier:  Essendant BOSGS20DT
Encompass_Preferred
Glue sticks are compatible with standard hot-melt low-temperature and dual-temperature glue guns providing versatile bonding performance for a variety of projects Excellent for dual- and low-temperature glue guns when applying to heat-sensitive materials Standard 4-inch glue sticks Designed specifically for use in Stanley GlueShot Hot Melt Glue Guns (GR20) and Stanley Dual Melt Glue Guns (BOS25-2) sold separately Adhesive Type- Solid Product Packaging- Stick Washable/Removable- Yes Color on Application- Clear
Catalog No. 50-000-22563