Learn More
Medchemexpress LLC HY-W019870 100mg Medchemexpress, Glufosinate (ammonium) CAS:77182-82-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW019870100MG
Encompass_Preferred
Medchemexpress, HY-W019870 100mg Glufosinate (ammonium) CAS:77182-82-2 Glufosinate ammonium, a phosphinic acid analogue of glutamic acid, is an herbicide which is converted by plant cells into PT (L-phosphinothricin). Glufosinate ammonium exerts neurotoxic activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0734