Learn More
Medchemexpress LLC Glufosinate ammonium | 77182-82-2 | MFCD00055562 | 99.9% | 198.16 g·mol-1 | C5H15N2O4P | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW019870R
Encompass_Preferred
Glufosinate ammonium is an analytical standard of a broad-spectrum herbicide used for research and method development. Supplied as a high-purity solid, it is intended for qualitative and quantitative analyses and is commonly used in HPLC, GC, and MS workflows for calibration, method validation, and residue analysis.
  • High purity suitable for analytical methods.
  • Intended for HPLC, GC, and MS applications.
  • Suitable for calibration and method validation.
  • Available in small pack sizes optimized for laboratory use.
  • Batch-specific documentation provided for traceability.
Catalog No. 50-003-02449