Learn More
Genetex Inc GluT1 Monoclonal Antibody [MSVA-401R] HistoMAX™ (GTX04469) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04469
Encompass_Preferred
The GluT1 antibody [MSVA-401R] HistoMAX™ (GTX04469) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human cerebrum (grey matter) tissue using GTX04469 GluT1 antibody [MSVA-401R] HistoMAX. A particularly strong GluT1 staining of endothelial cells is seen in the brain.
Catalog No. NC3667758