Learn More
Medchemexpress LLC Glutaconic acid | 1724-02-3 | MFCD00002784 | 99.6% | 130.10 g/mol | C5H6O4 | 100 MG

Supplier: Medchemexpress LLC HYW073501A100MG
Glutaconic acid is a small organic dicarboxylic acid provided for laboratory research use. It is supplied with supporting documentation and is intended for use as a reagent or intermediate in chemical research.
- High purity suitable for research applications.
- Molecular formula C5H6O4.
- Molecular weight 130.10 g/mol.
- Available in multiple pack sizes, including a 100 mg pack.
- Certificate of analysis and safety data sheet available.
- For research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.