Learn More
Medchemexpress LLC Glutaminase | 9001-47-2 | 10 U

Supplier: Medchemexpress LLC HYP276010U
Glutaminase is a metabolic enzyme that catalyzes the hydrolysis of glutamine to glutamate. It exists as isoenzymes encoded by Gls and Gls2 genes, with GLS and GLS2 proteins exhibiting distinct kinetic, immunologic, and molecular characteristics. This enzyme is utilized in cancer research.
- Catalyzes the hydrolysis of glutamine to glutamate.
- Used in cancer research.
- Available as an off-white to light yellow solid.
- Specific activity is 50-200 U/mg protein.
- One unit deaminates 1.0 μmole of L-glutamine per minute at pH 4.9 and 37°C.
- Soluble in H₂O at 1 mg/mL, requiring ultrasonic and warming.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.