Learn More
Medchemexpress LLC HY-114334 10mg Medchemexpress, Glutaminase-IN-1 CAS:2247127-79-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11433410MG
Encompass_Preferred
Medchemexpress, HY-114334 10mg Glutaminase-IN-1 CAS:2247127-79-1 Glutaminase-IN-1 (CB839 derivative), a CB839 derivative, is an allosteric inhibitor of 1,3,4-selenadiazole-containing kidney-type glutaminase (KGA), with an IC50 of 1 nM. Glutaminase-IN-1 (CB839 derivative) shows improved cellular uptake and antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9330