Learn More
Medchemexpress LLC Glutaminase-IN-3 | 1439399-45-7 | 99.9% | C19H19F3N6O2S | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY79583
Encompass_Preferred
Glutaminase-IN-3 (compound 657) is a potent glutaminase inhibitor with an IC50 of 0.24 μM for Glutaminase 1 (GLS1). It is extracted from patent WO2014089048A1, compound 657. This product is for research use only.
  • Potent glutaminase inhibitor
  • Inhibits Glutaminase 1 (GLS1) with an IC50 of 0.24 μM
  • High purity of 99.9%
  • Available as a white to yellow solid
  • Suitable for research applications
Catalog No. 50-003-58269