Learn More
Cayman Chemical GlutamInase-IN-3 250mg
SDP
Supplier:  Cayman Chemical 42325250 MG
Encompass_Preferred
An inhibitor of GAC (IC50 = 0.24 µM); inhibits the proliferation of PC3 and LNCaP prostate cancer cells (IC50s = 6.14 and 2.13 µM, respectively) and non-cancerous CCD-1072Sk fibroblasts (IC50 = 15.39 µM)
Catalog No. 50-294-3425