Learn More
Cayman Chemical GlutamInase InhIbItr comp 10mg
SDP
Supplier:  Cayman Chemical 1719910 MG
Encompass_Preferred
A cell-permeable, reversible inhibitor of a mitochondrial glutaminase splice variant, glutaminase C (IC50 = ~2.5 µM), which is commonly found in cancer cells; blocks transformation induced by Rho GTPases and inhibits the proliferation of cancer cell lines without affecting normal cells
Catalog No. 50-296-5441