Learn More
U.S. Pharmacopeia Glutamine, 56-85-9, MFCD00008044, 100mg
Supplier:  U.S. Pharmacopeia 1294808
Encompass
Molecular formula C5H10N2O3, Molecular weight 146.15, Melting Point 365 - 366.8 °F (185 - 186 °C) (decomposes)
Catalog No. NC0509492