Learn More
Medchemexpress LLC Glutamine Synthetase Antibody (YA751) | Protein G affinity purified | 42 kDa | 10 UL
SDP
Supplier:  Medchemexpress LLC HYP8040810UL
Encompass_Preferred
Glutamine Synthetase Antibody (YA751) is a Mouse-derived and non-conjugated IgG2a monoclonal antibody that targets Glutamine Synthetase (GLUL). This antibody is protein G affinity purified and has a molecular weight of 42 kDa. It is designed for research applications involving the study of GLUL, an enzyme crucial for ammonia regulation and detoxification in various biological processes.
  • Mouse-derived IgG2a monoclonal antibody
  • Targets Glutamine Synthetase (GLUL)
  • Protein G affinity purified
  • Molecular weight of 42 kDa
  • Supports research on ammonia regulation in the brain
  • Aids studies on ammonia removal in the liver
  • Useful for investigating ammonium detoxification during erythropoiesis
  • Contributes to ribosomal 40S subunit biogenesis research
  • Modulates BEST2 channel activity
Catalog No. 50-003-98940