Learn More
Genetex Inc Glutamine synthetase Monoclonal Antibody [MSVA-750M] HistoMAX™ (GTX04471) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04471
Encompass_Preferred
The Glutamine synthetase antibody [MSVA-750M] HistoMAX™ (GTX04471) is a Mouse Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human skin tissue using GTX04471 Glutamine synthetase antibody [MSVA-750M] HistoMAX. A nuclear and cytoplasmic glutamine synthetase staining can be seen in the lower third and in the top layers of the squamous epithelium.
Catalog No. NC3667762