Learn More
Medchemexpress LLC γ-Glutamylphenylalanine (Phe-13C9, 15N) | 7432-24-8 | 98.0% | 304.23 | C14H18N2O5 (13C9,15N-labeled) | 1 MG
SDP
Supplier:  Medchemexpress LLC HY101399SA1MG
Encompass_Preferred
γ-Glu-(Phe-13C9, 15N) is an isotopically labeled γ-glutamyl-phenylalanine supplied for analytical and research applications. It is intended for use as an internal standard or tracer in mass spectrometry and metabolic studies, enabling accurate quantitation and isotopic tracing while maintaining the chemical properties of the native dipeptide.
  • Isotope-labeled for mass spectrometry internal standard use.
  • High isotopic enrichment and analytical purity for reliable quantitation.
  • Chemical behavior mirrors the native dipeptide for accurate matrix matching.
  • Supplied as a stable dry powder suitable for storage and reconstitution.
  • Molecular weight 304.23 for reference in method development.
Catalog No. 50-002-13029