Learn More
Electron Microscopy Sciences Glutaraldehyde 0.625% in Phosphate Buffer, pH 7.4 500 ML
SDP
Supplier:  Electron Microscopy Sciences 1653505
Encompass_Preferred
Glutaraldehyde is available for your convenience in a concentration of 0.625% in Phosphate Buffer.
pH: 7.4
Catalog No. 50-366-946