Learn More
Medchemexpress LLC Glutaric acid-d6 | 154184-99-3 | 98.0% | 138.15 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW008820S5MG
Encompass_Preferred
Glutaric acid-d6 is the deuterium-labeled form of Glutaric acid. Glutaric acid is an intermediate in the catabolic pathways of lysine and tryptophan. It affects pericyte contractility and migration and is an indicator of glutaric aciduria type I. This compound is intended for research use only and is not sold to patients.
  • Can be utilized as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium substitution may impact pharmacokinetic and metabolic profiles.
Catalog No. 50-004-53071