Learn More
Medchemexpress LLC Glutarimide-isoindolinone-NH-PEG2-COOH | 2803827-03-2 | 98.5% | 419.43 | 100 MG
Supplier: Medchemexpress LLC HY138792100MG
Glutarimide-Isoindolinone-NH-PEG2-COOH is a synthesized E3 ligase ligand-linker conjugate that incorporates a cereblon ligand and a linker used in PROTAC technology.
- Synthesized E3 ligase ligand-linker conjugate
- Incorporates a cereblon ligand
- Utilizes a linker in PROTAC technology
- Exploits the intracellular ubiquitin-proteasome system
- Selectively degrades target proteins
- Targets Cereblon
- For research use only