Learn More
Medchemexpress LLC Glutathione arsenoxide TFA | 00-00-0 | 99.9% | C20H26AsF3N4O11S | 10MG
SDP
Supplier:  Medchemexpress LLC HY130588A10MG
Encompass_Preferred
Glutathione arsenoxide TFA is an arsenic-containing derivative of glutathione used as a research reagent and tumour metabolism inhibitor. It modulates mitochondrial function and is employed in cellular studies probing mitochondrial targets and cancer cell metabolism. Supplied as the trifluoroacetate (TFA) salt in small milligram quantities, it is intended for laboratory research use only.
  • Arsenic-containing glutathione derivative that modulates mitochondrial function.
  • Acts as a tumour metabolism inhibitor; used in cancer research applications.
  • Suitable for cellular and in vitro biochemical assays.
  • High purity (≈99.9%).
  • Available in small milligram quantities for exploratory studies.
Catalog No. 50-002-60932