Learn More
Cambridge Isotope Laboratories GLUTATHIONE -GSH- GLYCINE 50MG
Supplier:  Cambridge Isotope Laboratories CNLM624550
Encompass
Glutathione (GSH; 13C2/15N) is a stable isotope-labeled tripeptide used in MS-based proteomic methods for quantitative purposes. 50mg
Catalog No. NC3432808