Learn More
Genscript Corporation Glutathione MagBeads
Supplier:  Genscript Corporation L0089510
Encompass
The GenScript Glutathione MagBeads are developed for quick and efficient small scale purification of recombinant glutathione S transferase (GST)-tagged fusion proteins from bacteria yeast and mammalian crude cell lysates
Catalog No. 50-000-58652