Learn More
Bachem Glutathione-monoethyl ester (reduced)
SDP
Supplier:  Bachem 4015775.0250
Encompass_Preferred
Glutathione-monoethyl ester (reduced) , H-Glu(Cys-Gly-OEt)-OH, 92614-59-0, C12H21N3O6S, Mr 335.38, 250mg. Glutathione-monoethyl ester is effectively transported into many cell types including erythrocytes and converted intracellularly into glutathione. The compound may therefore be useful as a protective agent against the effects of radiation and other types of toxicity as well as in experiments on glutathione transport, metabolism, and function.
Catalog No. 50-259-485