Learn More
Biotium Glutathione S-Transferase Mu2 (GSTM2)(CPTC-GSTMu2-2), Biotin conjugate, 0.1mg/mL
Supplier:  Biotium BNCB2242500
Encompass_Preferred
Recognizes Glutathione S-Transferase of the mu class, which is involved in the detoxification of toxins, drugs, and products of oxidative stress by conjugation with glutathione. Primary antibodies are available purified, or with a selection of fluorescent CF Dyes and other labels. CF Dyes offer exceptional brightness and photostability. Note: Conjugates of blue fluorescent dyes like CF405S and CF405M are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors.
Catalog No. 50-006-90401