Learn More
Bioworld GLUTATHIONE SEPAROPORE SUPE
Supplier:  Bioworld 201810882
Encompass
GLUTATHIONE SEPAROPORE SUPER FLOW (SF): Used for protein purification by batch/column and scale-up procedures. Glutathione SeparoporeSuper Flow (SF) has an exellent protein binding capacity. The good flow properties of the 92 m (average ...
Catalog No. 50-197-895