Learn More
Sigma Aldrich Fine Chemicals Biosciences Glutathione SepharoseR Hig
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE17527901
Encompass
Glutathione Sepharose(R) High Performance is an affinity medium for high reSOLUTIONon purification of Glutathione S transferase (GST) tagged proteins.Glutathione Sepharose(R) High Performance is used for the purification of GST-tagged proteins from bacterial lysates and other S-transferases or glutathione-dependent proteins. GST-tagged proteins are eluted under mild nondenaturing conditions using reduced glutathione. The purification process preserves protein antigenicity and function.
Catalog No. 50-290-7591