Learn More
StatLab Glutaraldehyde 2% in 0.1M Phosphate Buffer pH 7.4 8oz (237mls)
SDP
Supplier:  StatLab S21588OZ
Encompass_Preferred
Glutaraldehyde 2% in 0.1M Phosphate Buffer pH 7.4 8oz (237mls)| EA | Maximum 6 months Refrigerate
Catalog No. NC0662486