Learn More
ORS Nasco GLV 11LT WT BUTYL RGH 10 1PR
Supplier:  ORS Nasco 068B131R/10
Encompass_Preferred
Honeywell part# B131R/9. Honeywell North 13 mil Butyl Chemical Resistant Gloves Black Glove Cuff Style: Beaded/Rolled Latex Free: Yes Size: 9 Style: North Butyl B131 Thickness: 13 mil 0.33 mm Certifications/Compliance: ASTM D3389-05 ASTM F790-97 OSHA 29 CFR EN 374 EN 420 EN388 Standard: EN 388 Hand: Specific
Catalog No. 50-255-1645