Learn More
ORS Nasco GLV BUTYL 13ML SZ 8 1PR
Supplier:  ORS Nasco 068B131/8
Encompass_Preferred
Honeywell part# B161/10. Honeywell North 16 mil Butyl Chemical Resistant Gloves Size: 10 Grip Design: Smooth Black Glove Cuff Style: Beaded/Rolled Latex Free: Yes Style: North Butyl B161 Thickness: 16 mil 0.40 mm Certifications/Compliance: ASTM D3389-05 ASTM F790-97 OSHA 29 CFR EN 374 EN 420 Hand: Specific
Catalog No. 50-255-1647