Learn More
Medchemexpress LLC Gly-7-MAD-MDCPT hydrochloride | 98.3% | 514.92 | C24H23ClN4O7 | 5 MG

Supplier: Medchemexpress LLC HY132164A5MG
Gly-7-MAD-MDCPT hydrochloride is a camptothecin-derived anticancer payload supplied as the hydrochloride salt for research use. It is provided as a high-purity material suitable for antibody-drug conjugate development and in vitro cytotoxicity testing.
- High purity of 98.3% for research-grade applications.
- Hydrochloride salt form to improve solubility and handling.
- Molecular weight 514.92 g/mol.
- Chemical formula C24H23ClN4O7.
- Available in small research pack sizes (for example, 5 mg).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.