Learn More
Medchemexpress LLC Gly-Dasatinib | 2102305-55-3 | 501.00 | 25 MG

Supplier: Medchemexpress LLC 25MGHY176462
Gly-Dasatinib (compound 1-7-1), an amino acid analog of Dasatinib (HY-10181), is a tyrosine kinase inhibitor. It can be used for the study of cancer. Gly-Dasatinib is a specialized tyrosine kinase inhibitor, functioning as an amino acid analog of Dasatinib. This product is intended for research purposes, specifically in the study of cancer. It is categorized as a drug derivative and should be stored under recommended conditions as specified in its Certificate of Analysis.
- Target: Drug Derivative
- Pathway: Others
- Application: Can be used for the study of cancer
- Classification: Tyrosine kinase inhibitor
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.