Learn More
Medchemexpress LLC Gly-gly-gly-PEG4-azide | 3070260-22-6 | 99.8% | 433.46 | C16H31N7O7 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY14506610MG
Encompass_Preferred
Gly-Gly-Gly-PEG4-azide is a cleavable azide-functional linker used in antibody-drug conjugate (ADC) synthesis and bioconjugation. It provides an azide handle for copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted alkyne-azide cycloaddition (SPAAC), enabling bioorthogonal conjugation of payloads to antibodies and other biomolecules.
  • Contains an azide functional group for bioorthogonal click reactions.
  • Cleavable linker compatible with ADC synthesis workflows.
  • High purity suitable for research applications.
  • Oil appearance with defined solvent storage stability under nitrogen.
  • Available in small-scale milligram pack sizes for research use.
Catalog No. 50-002-03566