Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gly-gly-gly-PEG4-DBCO | 2353409-80-8 | 95.1% | 694.77 g/mol | C35H46N6O9 | 100 MG
SDP

Supplier:  Medchemexpress LLC HY140309100MG

Encompass_Preferred

Gly-gly-gly-PEG4-DBCO is a cleavable 4-unit polyethylene glycol (PEG4) linker bearing a DBCO (dibenzocyclooctyne) moiety for strain-promoted azide-alkyne cycloaddition. It is supplied as a dry powder intended for bioconjugation and antibody-drug conjugate (ADC) synthesis.

  • Cleavable 4-unit PEG linker suitable for ADC synthesis.
  • Contains a DBCO handle for copper-free click chemistry (SPAAC).
  • High purity (95.1%) and molecular weight of 694.77 g/mol.
  • Soluble in DMSO; ultrasonic assistance recommended for dissolution.
  • Powder stable at -20°C; store solutions at -80°C for long-term stability.

Catalog No. 50-002-02676


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.