Learn More
Medchemexpress LLC Gly-Gly-Phe-Gly-NH-CH2-O-CH2COOH | 1599440-20-6 | 99.9% | C18H25N5O7 | 100 MG

Supplier: Medchemexpress LLC HY45672100MG
Gly-Gly-Phe-Gly-NH-CH2-O-CH2COOH is an ADC Linker that can be used to synthesize Drug-Linker Conjugates for ADC. It is especially used for the synthesis of Deruxtecan (HY-13631E), which is a toxic drug-linker conjugate. This product is for research use only and is not sold to patients.
- Used to synthesize drug-linker conjugates for ADC
- Specifically for the synthesis of Deruxtecan, a toxic drug-linker conjugate
- Molecular weight: 423.42
- Appearance: solid
- Color: white to off-white
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.