Learn More
Medchemexpress LLC Gly-gly-phe-gly-nh-o-co-exatecan | 1599440-12-6 | 95.9% | 840.85 | C42H45FN8O10 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY13631M5MG
Encompass_Preferred
Gly-Gly-Phe-Gly-NH-O-CO-Exatecan is a drug-linker conjugate intended for antibody-drug conjugate (ADC) research. It couples a cleavable Gly-Gly-Phe-Gly peptide linker to the topoisomerase I inhibitor exatecan, enabling targeted delivery of a cytotoxic payload. Supplied as an off-white to light yellow solid with supporting COA and handling documentation for research use.
Catalog No. 50-002-22496