Learn More
Medchemexpress LLC HY-114392 10mg Medchemexpress, Gly-β-MCA CAS:66225-78-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11439210MG
Encompass_Preferred
Medchemexpress, HY-114392 10mg Gly-β-MCA CAS:66225-78-3 Gly-β-MCA, a bile acid, is a potent, sable, intestine-selective and oral bioactive farnesoid X receptor (FXR) inhibitor that may be a candidate for the treatment of metabolic disorders. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9340