Learn More
Medchemexpress LLC Gly-Pro-pNA (hydrochloride) | 103213-34-9 | 99.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11924410MM 1ML
Encompass_Preferred
Gly-Pro-pNA hydrochloride is a chromogenic dipeptidyl peptidase IV (DPP-IV) substrate for enzymatic assays; cleavage releases p-nitroaniline (pNA), producing a measurable colorimetric signal useful for inhibitor screening and activity quantification. Supplied as a 10 mM solution option in DMSO, it is suitable for assay development and routine enzyme screening.
  • Chromogenic substrate for DPP-IV assays.
  • Releases p-nitroaniline for spectrophotometric detection.
  • High purity for reproducible assay results.
  • Provided as a 10 mM solution in DMSO, 1 mL vial.
  • Available as a solid for long-term storage.
  • Compatible with standard assay workflows and inhibitor screening.
Catalog No. 50-002-33030