Learn More
Medchemexpress LLC Gly-Pro-pNA (hydrochloride) | 103213-34-9 | 99.9% | 5 MG

Supplier: Medchemexpress LLC HY1192445MG
Gly-Pro-pNA hydrochloride is a chromogenic substrate for dipeptidyl peptidase IV (DPP-IV) that releases p-nitroaniline upon enzymatic cleavage. It is used in enzymatic assays to screen for DPP-IV inhibitors and for experimental antidiabetic research. For research use only; not for human or veterinary use.
- Chromogenic substrate for DPP-IV assays.
- High purity (99.92%).
- Molecular formula C13H17ClN4O4; molecular weight 328.76.
- Solid, white to yellow appearance.
- Soluble in DMSO (100 mg/mL) and water (50 mg/mL); may require ultrasonic.
- Recommended storage sealed at 4°C; in solution store -80°C (6 months) or -20°C (1 month).
- Available in small pack sizes including 5 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.