Learn More
Medchemexpress LLC Glyasperin A | 142474-52-0 | 1 MG

Supplier: Medchemexpress LLC HYN39561MG
Glyasperin A is a flavonoid compound isolated from the acetone extract of the leaves of Macaranga gigantea.
- Flavonoid compound.
- Isolated from the acetone extract of the leaves of a plant called Macaranga gigantea.
- Intended for research use only.
- Has a molecular weight of 422.47.
- Has a chemical formula of C25H26O6.
- Appears as a light yellow to yellow solid.
- Classified as a flavonoid and flavonol.
- Has a purity of 99.65%.
- Can be stored at 4°C, protected from light.
- In solvent, can be stored at -80°C for 6 months or -20°C for 1 month, protected from light.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.