Learn More
Cayman Chemical GlyburIde-d3 1mg
SDP
Supplier:  Cayman Chemical 257401 MG
Encompass_Preferred
An internal standard for the quantification of glyburide by GC- or LC-MS
Catalog No. 50-293-6351