Learn More
NANOMICRO TECHNOLOGIES INC NanoChrom Technologies, BioCore Glycan LC Analytical Column, 1.8um, 2.1 x 150 mm, Stainless Steel
Supplier:  NANOMICRO TECHNOLOGIES INC B91301801802115S
Encompass_Preferred
BioCore Glycan is a family of high-performance HILIC columns designed for profiling N-glycans present in proteins and related substances. This column technology involves the creation of a proprietary hydrophilic layer on the surface of monodispersed, spherical, porous silica particles, combined with well-established column packing processes.
Catalog No. NC2815337