Learn More
Bioss Glyceraldehyde 3-Phosphate Deh
SDP
Supplier:  Bioss BSM70126M
Encompass_Preferred
Glyceraldehyde 3-Phosphate Dehydrogenase Primary Antibody Unconjugated Host Species Mouse Target Species Human Mouse Rat Bovine Clonality Monoclonal Applications WB IHC 1D4Concentration Lot Dependent Size 100 ul Storage Buffer PBS 50% glycerol and 5 mm NaN3
Catalog No. 50-000-65117