Learn More
Industrial Chemical & Scientific Co Glycerin, MFCD00004722, 1Gallon
Supplier:  Industrial Chemical & Scientific Co 86041
Encompass_Preferred
Molecular Formula C3H8O3, Purity 99.5%, Molecular Weight 92.093
Catalog No. NC9234020