Learn More
Medchemexpress LLC Glycerol-13C-1 | 82425-96-5 | 99.3% | 93.09 g/mol | C3H8O3 (1-13C) | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB1659S210MG
Encompass_Preferred
Glycerol-13C-1 (90% in water) is a 13C-labeled glycerol solution supplied as a liquid analytical reagent for use as a tracer and internal standard in quantitative analyses. It is commonly used in sample preparation and polyacrylamide gel electrophoresis, and is suitable for NMR, GC-MS, and LC-MS applications. Documents such as COA and SDS are available.
Catalog No. 50-002-97241