Learn More
Medchemexpress LLC Glycerol-13C2 (Trihydroxypropane-13C2) | 102088-01-7 | 99% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYB1659S4
Encompass_Preferred
Glycerol-13C2 is the 13C labeled Glycerol. Glycerol is utilized in sample preparation and gel formation for polyacrylamide gel electrophoresis. It can also be used as a tracer and an internal standard for quantitative analysis through methods like NMR, GC-MS, or LC-MS.
  • used in sample preparation for polyacrylamide gel electrophoresis
  • utilized for gel formation in polyacrylamide gel electrophoresis
  • can be applied as a tracer
  • functions as an internal standard for quantitative analysis by NMR
  • serves as an internal standard for quantitative analysis by GC-MS
  • acts as an internal standard for quantitative analysis by LC-MS
Catalog No. 50-004-14334