Learn More
Medchemexpress LLC Glycerol-3-phosphate oxidase, microorganism | 9046-28-0 | 1 KU
SDP
Supplier:  Medchemexpress LLC HYP2733C1KU
Encompass_Preferred
Glycerol-3-phosphate oxidase, microorganism, is an enzyme that catalyzes the oxidation of glycerol-3-phosphate to dihydroxyacetone phosphate, producing hydrogen peroxide. It is located in the mitochondria of microorganisms, where it participates in the glycerol-3-phosphate cycle, regulating cellular energy metabolism.
  • Catalyzes the oxidation of glycerol-3-phosphate
  • Produces hydrogen peroxide
  • Regulates cellular energy metabolism
  • Can be used with Lipoprotein Lipase and Glycerol Kinase to determine triglyceride levels
Catalog No. 50-004-00269