Learn More
Medchemexpress LLC Glycerol | 56-81-5 | 98.91% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB1659R500MG
Encompass_Preferred
Glycerol (Standard) is the analytical standard of Glycerol, intended for research and analytical applications. It is used in sample preparation and gel formation for polyacrylamide gel electrophoresis. This analytical standard is a reference standard supplied for assay and is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for research
  • Used in sample preparation and gel formation
  • Reference standard for assays
  • Suitable for HPLC, GC, and MS research
Catalog No. 50-004-13234