Learn More
Alkali Scientific Glycerol, ACS Grade, 1 L
Supplier: Alkali Scientific LYC005CP
Glycerol, ACS Grade, 1 L, is a high-purity, versatile laboratory reagent commonly used in a variety of applications including cryopreservation, biochemical research, and chemical synthesis. It is frequently used as a humectant, helping to retain moisture in both pharmaceutical and cosmetic formulations. Glycerol is also vital in cryopreservation of cells, tissues, and other biological materials by preventing the formation of ice crystals during freezing. This product meets the American Chemical Society (ACS) grade standards, ensuring its high purity and suitability for use in scientific experiments and pharmaceutical preparations. It is widely used in both academic research and industrial applications.