Learn More
Alkali Scientific Glycerol, ACS Grade, 4 L
Supplier: Alkali Scientific LYC008CP
Glycerol, ACS Grade, 4 L, is a high-purity laboratory-grade reagent used in a wide range of applications including cryopreservation, chemical synthesis, and buffer preparation. It serves as a humectant, helping to retain moisture in various formulations, such as skin care products, ointments, and pharmaceutical solutions. Glycerol is also crucial in biological research for the preservation of cells and tissues, preventing ice damage during freezing. With its high purity, this ACS-grade glycerol is ideal for use in sensitive laboratory procedures, ensuring minimal interference in chemical and biochemical experiments. It is widely used in both academic and industrial research settings.