Learn More
Medchemexpress LLC Glycerol-d3 | 7325-16-8 | >98% | 500 MG
Supplier: Medchemexpress LLC HYB1659S8500MG
Glycerol-d3 is the deuterium labeled Glycerol. Glycerol is utilized in sample preparation and gel formation for polyacrylamide gel electrophoresis. It can be used as a tracer and as an internal standard for quantitative analysis through NMR, GC-MS, or LC-MS. Deuteration of drug molecules, which involves incorporating stable heavy isotopes of hydrogen, carbon, and other elements, can impact their pharmacokinetic and metabolic profiles.
- Used in sample preparation and gel formation for polyacrylamide gel electrophoresis.
- Can be used as a tracer and internal standard for quantitative analysis.
- Useful for NMR, GC-MS, or LC-MS applications.
- Deuteration can impact pharmacokinetic and metabolic profiles of drug molecules.